2-Chloropyridine, 99%, Thermo Scientific Chemicals
2-Chloropyridine, 99%, Thermo Scientific Chemicals
2-Chloropyridine, 99%, Thermo Scientific Chemicals
2-Chloropyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloropyridine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR110011000
Price (MYR)
427.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
427.00
Add to cart
Chemical Identifiers
CAS673-06-3
IUPAC Name(2R)-2-amino-3-phenylpropanoic acid
Molecular FormulaC9H11NO2
InChI KeyCOLNVLDHVKWLRT-MRVPVSSYSA-N
SMILESN[C@H](CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Loss on drying=<0.5 %
Sulfated ash=<0.1 %
Heavy metals (as Pb)=<20 ppm