2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(4-Chlorophenyl)ethylamine, 98%, Thermo Scientific Chemicals
CAS: 156-41-2 | C8H11ClN | 156.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number ACR109880250
Price (MYR)
813.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
813.00
Add to cart
Specifications
Chemical Name or Material2-(4-Chlorophenyl)ethylamine
Name Note98%
CAS156-41-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger