4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
CAS: 604-44-4 | C10H7ClO | 178.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR109551000
Price (MYR)
3,421.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,421.00
Add to cart
Specifications
Chemical Name or Material4-Chloro-1-naphthol
Name Note99%
CAS604-44-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning