4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-1-naphthol, 97%, Thermo Scientific Chemicals
CAS: 604-44-4 | C10H7ClO | 178.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR109550250
Price (MYR)
1,096.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,096.00
Add to cart
Chemical Identifiers
CAS52488-36-5
IUPAC Name4-bromo-1H-indole
Molecular FormulaC8H6BrN
InChI KeyGRJZJFUBQYULKL-UHFFFAOYSA-N
SMILESBrC1=C2C=CNC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.6520-1.6610 @20?C
Appearance (Color)Yellow to dark brown
Assay (GC)≥95.0%