2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, Thermo Scientific Chemicals
2-Chloro-2-methylpropane, 99%, C4H9Cl, CAS Number-507-20-0, 2-methyl-2-chloropropane, propane, 2-chloro-2-methyl, 2-chloroisobutane, tert-butylchloride, t-butylchloride, chlorotrimethylmethane, trimethylchloromethane, tert-butyl chloride, t-butyl chloride, 2-methyl-2-propyl chloride, 500mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
Packaging:
Glass bottle
Catalog number ACR109505000
Price (MYR)
559.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
559.00
Add to cart
Chemical Identifiers
CAS39409-82-0
IUPAC Namepentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
Molecular FormulaC4H10Mg5O18
InChI KeyYMKSTRLFWJQKFP-UHFFFAOYSA-D
SMILESO.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.