4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Catalog number ACR109485000
Price (MYR)
651.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
651.00
Add to cart
Chemical Identifiers
CAS13930-88-6
IUPAC Name2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide; oxovanadiumbis(ylium)
Molecular FormulaC32H16N8OV
InChI KeyYRZZLAGRKZIJJI-UHFFFAOYSA-N
SMILESO=[V++].[N-]1C2=NC3=C4C=CC=CC4=C([N-]3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to purple
FormPowder
Assay from Suppliers CofA≥80.0%