4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Catalog number ACR109481000
Price (MYR)
427.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
427.00
Add to cart
Chemical Identifiers
CAS389-08-2
IUPAC Name1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Molecular FormulaC12H12N2O3
InChI KeyMHWLWQUZZRMNGJ-UHFFFAOYSA-N
SMILESCCN1C=C(C(O)=O)C(=O)C2=CC=C(C)N=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration with NaOH>=99.4 %
Appearance (Form)Powder
Melting point225°C to 231°C