4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Catalog number ACR109480050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Chloro-3-methylphenol
Name Note99+%
CAS59-50-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger