5-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
5-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
5-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
5-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
CAS: 95-79-4 | C7H8ClN | 141.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR109380050
Price (MYR)
380.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
380.00
Add to cart
Chemical Identifiers
CAS2206-27-1
IUPAC Name(²H₃)methanesulfinyl(²H₃)methane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.75 %
Water=<0.03 % (Coulometric)
TMS0.02 to 0.04 v/v %