4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number ACR109371000
Price (MYR)
5,103.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,103.00
Add to cart
Specifications
Chemical Name or Material4-Chloro-2-methylaniline
Name Note98%
CAS95-69-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger