4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-2-methylaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR109370050
Price (MYR)
594.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
594.00
Add to cart
Chemical Identifiers
CAS7440-64-4
IUPAC Nameytterbium
Molecular FormulaYb
InChI KeyNAWDYIZEMPQZHO-UHFFFAOYSA-N
SMILES[Yb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.