5-Chloroindole, 99%, Thermo Scientific Chemicals
5-Chloroindole, 99%, Thermo Scientific Chemicals
5-Chloroindole, 99%, Thermo Scientific Chemicals
5-Chloroindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chloroindole, 99%, Thermo Scientific Chemicals
CAS: 17422-32-1 | C8H6ClN | 151.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR109310010
Price (MYR)
592.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
592.00
Add to cart
Specifications
Chemical Name or Material5-Chloroindole
Name Note99%
CAS17422-32-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning