1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 2008-75-5 | C7H14ClN·HCl | 184.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR109211000
Price (MYR)
522.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
522.00
Add to cart
Chemical Identifiers
CAS1532-71-4
IUPAC Name1-bromoisoquinoline
Molecular FormulaC9H6BrN
InChI KeyYWWZASFPWWPUBN-UHFFFAOYSA-N
SMILESBrC1=C2C=CC=CC2=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point42°C to 48°C
Appearance (Color)Yellow to brown
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
HPLC>=95.0 %