4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 88-04-0 | C8H9ClO | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR109065000
Price (MYR)
713.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
713.00
Add to cart
Chemical Identifiers
CAS593-84-0
IUPAC Nameguanidine; sulfanylformonitrile
Molecular FormulaC2H6N4S
InChI KeyZJYYHGLJYGJLLN-UHFFFAOYSA-N
SMILESSC#N.NC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or pale pink
Assay (Argentometric Titration)≥98.5 to ≤101.5% (non-U.S. sourced material)
Identification (FTIR)Conforms (non-U.S. sourced material)
FormCrystalline powder
CommentMaterial sourced in the U.S. and in other countries