4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 88-04-0 | C8H9ClO | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR109060025
Price (MYR)
2,369.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,369.00
Add to cart
Chemical Identifiers
CAS7447-41-8
IUPAC Namelithium(1+) chloride
Molecular FormulaClLi
InChI KeyKWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES[Li+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or granules
Sulfate (SO4)=<0.01 %
Potassium (K)=<0.0015 %
Appearance (Color)White
Titration Argentometric98.5 to 101.5 %