2-Chlorobutane, 99+%, Thermo Scientific Chemicals
2-Chlorobutane, 99+%, Thermo Scientific Chemicals
2-Chlorobutane, 99+%, Thermo Scientific Chemicals
2-Chlorobutane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorobutane, 99+%, Thermo Scientific Chemicals
CAS: 78-86-4 | C4H9Cl | 92.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number ACR108832500
Price (MYR)
489.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
489.00
Add to cart
Chemical Identifiers
CAS827614-66-4
IUPAC Name1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular FormulaC13H23BN2O2
InChI KeyYMEBZRNYQBODKB-UHFFFAOYSA-N
SMILESCC(C)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid