3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
CAS: 618-46-2 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number ACR108740050
Price (MYR)
394.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Chemical Identifiers
CAS52093-26-2
IUPAC Namelanthanum(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9LaO9S3
InChI KeyWGJJZRVGLPOKQT-UHFFFAOYSA-K
SMILES[La+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder or crystals
Elemental Analysis23.35 to 24.05 % (La)
Appearance (Color)White to light beige