4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number ACR108591000
Price (MYR)
415.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
415.00
Add to cart
Chemical Identifiers
CAS688-73-3
IUPAC Nametributylstannyl
Molecular FormulaC12H27Sn
InChI KeyPIILXFBHQILWPS-UHFFFAOYSA-N
SMILESCCCC[Sn](CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4720 to 1.4740 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %