4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number ACR108590050
Price (MYR)
428.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
428.00
Add to cart
Chemical Identifiers
CAS13454-96-1
IUPAC Nametetrachloroplatinum
Molecular FormulaCl4Pt
InChI KeyFBEIPJNQGITEBL-UHFFFAOYSA-J
SMILESCl[Pt](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<25 ppm
Silicon (Si)=<200 ppm
Trace metal analysis=<500 ppm (Sum Ru+Rh+Ir+Pd)
Appearance (Color)Brown
Iron (Fe)=<50 ppm