4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR108590010
Price (MYR)
1,194.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,194.00
Add to cart
Chemical Identifiers
CAS7439-95-4
IUPAC Namemagnesium
Molecular FormulaMg
InChI KeyJLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver white
Total trace metal impurities=<0.10 %
Assay>=99.90 %
Total trace metal impuritiesmetals sought for: Si, Mn, Cu, Al, Fe, Pb, Ni, Sn
Appearance (Form)Turnings