4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
CAS: 99-91-2 | C8H7ClO | 154.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
Glass bottle
Catalog number ACR108542500
Price (MYR)
646.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
646.00
Add to cart
Chemical Identifiers
CAS7440-67-7
IUPAC Namezirconium
Molecular FormulaZr
InChI KeyQCWXUUIWCKQGHC-UHFFFAOYSA-N
SMILES[Zr]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size<45?m (325 mesh): ≥99.9%
Impurity contentHafnium: ca. 2%
Particle SizeAverage: 4.4-6.5?m
Assay (unspecified)Zirconium (incl. Hf) total: ≥98.8%