Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
CAS: 97-97-2 | C4H9ClO2 | 124.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number ACR108500010
Price (MYR)
725.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
725.00
Add to cart
Specifications
Chemical Name or MaterialChloroacetaldehyde dimethyl acetal
Name Note97%
CAS97-97-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.