Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
CAS: 86-74-8 | C12H9N | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
25 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number ACR108262500
Price (MYR)
831.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
831.00
Add to cart
Specifications
Chemical Name or MaterialCarbazole
Name Note96%
CAS86-74-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Suspected of causing cancer.