Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
CAS: 86-74-8 | C12H9N | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number ACR108260250
Price (MYR)
467.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
467.00
Add to cart
Chemical Identifiers
CAS112734-22-2
IUPAC Name1-(4-bromo-2-fluorophenyl)methanamine
Molecular FormulaC7H7BrFN
InChI KeyRLTFBWCBGIZCDQ-UHFFFAOYSA-N
SMILESNCC1=C(F)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5630 to 1.5660 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %