Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
CAS: 86-74-8 | C12H9N | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
250 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number ACR108260050
Price (MYR)
446.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
446.00
Add to cart
Chemical Identifiers
CAS50-01-1
IUPAC Namehydrogen guanidine chloride
Molecular FormulaCH6ClN3
InChI KeyPJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Water=<0.3 %
Anticaking agent=<0.4 % (SiO2)
Appearance (Color)White to light yellow
Melting point179°C to 186°C