Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
CAS: 86-74-8 | C12H9N | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
25 g
250 g
Packaging:
View all
Plastic bottle
Catalog number ACR108260010
Price (MYR)
2,110.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,110.00
Add to cart
Chemical Identifiers
CAS1141-88-4
IUPAC Name2-[(2-aminophenyl)disulfanyl]aniline
Molecular FormulaC12H12N2S2
InChI KeyYYYOQURZQWIILK-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1SSC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%