ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
CAS: 105-60-2 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR108231000
Price (MYR)
329.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
329.00
Add to cart
Chemical Identifiers
CAS930-68-7
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.4855-1.4905 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%