ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
CAS: 105-60-2 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR108230025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS59-92-7
IUPAC Name(2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
Molecular FormulaC9H11NO4
InChI KeyWTDRDQBEARUVNC-LURJTMIESA-N
SMILESN[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 % (On dried substance)
Appearance (Color)White to cream
Infrared spectrumConforms
Loss on drying=<1 %
Appearance (Form)Crystalline powder