ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
CAS: 105-60-2 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
250 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR108230010
Price (MYR)
507.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
507.00
Add to cart
Chemical Identifiers
CAS673-32-5
IUPAC Name(prop-1-yn-1-yl)benzene
Molecular FormulaC9H8
InChI KeyGHUURDQYRGVEHX-UHFFFAOYSA-N
SMILESCC#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5620-1.5660 @ 20?C