Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
The crystalline powder form of the stimulant caffeine, CAS # 58-08-2, purified for analytical chemistry applications such as HPLC.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR108165000
Price (MYR)
779.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
779.00
Add to cart
Specifications
Chemical Name or MaterialCaffeine
Name Note98.5%
Acidity0.2mL of 0.01M NaOH max.
CAS58-08-2
Health Hazard 1GHS Signal Word: Warning
View more
General description
  • Caffeine is an alkaloid vagal/vasomotor stimulant found in certain nuts, seeds, and legumes
  • It is a methylxanthine, derived from a purine base compound found in the tissues of many organisms
  • Caffeine competitively binds adenosine receptors and prevents signaling from dampening, thereby sustaining activation, and producing increases in energy
RUO – Research Use Only