Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Caffeine, 98.5%, specified according to the req. of USP/BP, Thermo Scientific Chemicals
The crystalline powder form of the stimulant caffeine, CAS # 58-08-2, purified for analytical chemistry applications such as HPLC.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
10 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR108160025
Price (MYR)
3,113.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,113.00
Add to cart
Chemical Identifiers
CAS7632-00-0
IUPAC Namesodium nitrite
Molecular FormulaNNaO2
InChI KeyLPXPTNMVRIOKMN-UHFFFAOYSA-M
SMILES[Na+].[O-]N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with KMnO4>=97.0 %
Heavy metals (as Pb)=<10 ppm
Insoluble matter=<0.01 %
Chloride (Cl)=<50 ppm
View more
General description
  • Caffeine is an alkaloid vagal/vasomotor stimulant found in certain nuts, seeds, and legumes
  • It is a methylxanthine, derived from a purine base compound found in the tissues of many organisms
  • Caffeine competitively binds adenosine receptors and prevents signaling from dampening, thereby sustaining activation, and producing increases in energy
RUO – Research Use Only