S-Butyrylthiocholine iodide, 98%, Thermo Scientific Chemicals
S-Butyrylthiocholine iodide, 98%, Thermo Scientific Chemicals
S-Butyrylthiocholine iodide, 98%, Thermo Scientific Chemicals
S-Butyrylthiocholine iodide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
S-Butyrylthiocholine iodide, 98%, Thermo Scientific Chemicals
CAS: 1866-16-6 | C9H20INOS | 317.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR108150250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS367-57-7
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear light yellow to orange
Appearance (Form)Liquid
AppearanceMay darken on storage
GC>=97.5 %