gamma-Butyrolactone, 99+%
gamma-Butyrolactone, 99+%
gamma-Butyrolactone, 99+%
gamma-Butyrolactone, 99+%
Thermo Scientific Chemicals
gamma-Butyrolactone, 99+%
CAS: 96-48-0 | C4H6O2 | 86.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR10813010010 LPlastic drum
Catalog number ACR108130100
Price (MYR)
3,267.00
Add to cart
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
3,267.00
Add to cart
Chemical Identifiers
CAS1008-88-4
IUPAC Name3-phenylpyridine
Molecular FormulaC11H9N
InChI KeyHJKGBRPNSJADMB-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.6135-1.6195 @ 20?C
Assay (GC)≥94.0%