gamma-Butyrolactone, 99+, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals
gamma-Butyrolactone, 99+, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR108130010
Price (MYR)
933.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
933.00
Add to cart
Chemical Identifiers
CAS107-91-5
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)118-123?C
Assay (GC)≥98.5%
Appearance (Color)White to pale cream