n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
CAS: 107-92-6 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
Glass bottle
Catalog number ACR108110025
Price (MYR)
672.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
672.00
Add to cart
Chemical Identifiers
CAS62-55-5
IUPAC Nameethanethioamide
Molecular FormulaC2H5NS
InChI KeyYUKQRDCYNOVPGJ-UHFFFAOYSA-N
SMILESCC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>/+ 99.0
CommentClarity of a 2% solution P.T.
Residue on ignition0.05%
Melting Point111-114?