n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
CAS: 107-92-6 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR108110010
Price (MYR)
546.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
546.00
Add to cart
Chemical Identifiers
CAS2975-46-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow to pale brown
Assay (unspecified)> 96.0%