n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR108015000
Price (MYR)
1,383.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,383.00
Add to cart
Chemical Identifiers
CAS96-31-1
IUPAC Name1,3-dimethylurea
Molecular FormulaC3H8N2O
InChI KeyMGJKQDOBUOMPEZ-UHFFFAOYSA-N
SMILESCNC(=O)NC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)White
Melting point103°C to 108°C
Appearance (Form)Crystalline powder or crystals