n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR108011000
Price (MYR)
577.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
577.00
Add to cart
Chemical Identifiers
CAS5856-63-3
IUPAC Name2-aminobutan-1-ol
Molecular FormulaC4H11NO
InChI KeyJCBPETKZIGVZRE-UHFFFAOYNA-N
SMILESCCC(N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid or viscous liquid
Optical Rotation-10.0 ± 0.1? (neat)
Refractive Index1.4505-1.4545 @ 20?C
Assay (GC)≥97.5%