4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
CAS: 98-54-4 | C10H14O | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
Catalog number ACR108000050
Price (MYR)
334.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
334.00
Add to cart
Chemical Identifiers
CAS298-14-6
IUPAC Namepotassium hydrogen carbonate
Molecular FormulaCHKO3
InChI KeyTYJJADVDDVDEDZ-UHFFFAOYSA-M
SMILES[K+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfur compounds (as SO4)=<0.003 %
Magnesium (Mg)=<0.001 %
Appearance (Color)Colorless to white
Titration with HCl99.7 to 100.5 % (On dried basis)
Phosphate (PO4)=<5 ppm