4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
CAS: 98-54-4 | C10H14O | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
Catalog number ACR108000010
Price (MYR)
585.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
585.00
Add to cart
Specifications
Chemical Name or Material4-tert-Butylphenol
Name Note97%
CAS98-54-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger