n-Butyl disulfide, 97%, Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
CAS: 629-45-8 | C8H18S2 | 178.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR107921000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104163-35-1
IUPAC Name1-(3-methylthiophen-2-yl)methanamine
Molecular FormulaC6H9NS
InChI KeySWZNXCABBUKIPZ-UHFFFAOYSA-N
SMILESCC1=C(CN)SC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥97.5%
Refractive Index1.5560-1.5600 @ 20?C