n-Butyl disulfide, 97%, Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
CAS: 629-45-8 | C8H18S2 | 178.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR107920250
Price (MYR)
464.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
464.00
Add to cart
Specifications
Chemical Name or Materialn-Butyl disulfide
CAS629-45-8
Health Hazard 1GHS Signal Word: Warning
ColorBrown to Colorless
Density0.9380g/mL