4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
CAS: 98-52-2 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
View all
Plastic bottle
Catalog number ACR107902500
Price (MYR)
682.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
682.00
Add to cart
Chemical Identifiers
CAS57-88-5
IUPAC Name(1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular FormulaC27H46O
InChI KeyHVYWMOMLDIMFJA-DPAQBDIFSA-N
SMILESCC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)White
Infrared spectrumConforms
Stabilizer50 to 350 ppm alpha-Tocopherol
Appearance (Form)Crystalline powder