4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
CAS: 98-52-2 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
Packaging:
View all
Glass Bottle
Catalog number ACR107900050
Price (MYR)
344.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
344.00
Add to cart
Specifications
Chemical Name or Material4-tert-Butylcyclohexanol
Name Note99%
CAS98-52-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning