tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl carbazate, 99%, Thermo Scientific Chemicals
CAS: 870-46-2 | C5H12N2O2 | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR107880250
Price (MYR)
604.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
604.00
Add to cart
Chemical Identifiers
CAS16937-99-8
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid hydrate
Molecular FormulaC11H23NO5
InChI KeyURQQEIOTRWJXBA-UHFFFAOYNA-N
SMILESO.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)3.75-10.46% (0.5-1.5 waters)
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Optical Rotation25.2 ± 0.1? (C=2 in acetic acid)
Appearance (Color)White