tert-Butyl acetate, 99%, Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
CAS: 540-88-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR107765000
Price (MYR)
1,286.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,286.00
Add to cart
Specifications
Chemical Name or Materialtert-Butyl acetate
Name Note99+%
CAS540-88-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Repeated exposure may cause skin dryness or cracking.