tert-Butyl acetate, 99%, Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
CAS: 540-88-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass bottle
Catalog number ACR107760025
Price (MYR)
3,700.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,700.00
Add to cart
Chemical Identifiers
CAS91-40-7
IUPAC Name2-(phenylamino)benzoic acid
Molecular FormulaC13H11NO2
InChI KeyZWJINEZUASEZBH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point182°C to 187°C
Appearance (Color)Yellow-green to grey-green
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Form)Powder