sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number ACR107700100
Price (MYR)
1,470.00
Add to cart
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
1,470.00
Add to cart
Chemical Identifiers
CAS78-92-2
IUPAC Namebutan-2-ol
Molecular FormulaC4H10O
InChI KeyBTANRVKWQNVYAZ-UHFFFAOYNA-N
SMILESCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.5 %
Color scale=<10 APHA
GC>=98.5 %