sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass Bottle
Catalog number ACR107700025
Price (MYR)
761.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
761.00
Add to cart
Specifications
Chemical Name or Materialsec-Butanol
Melting Point-115.0°C
CAS78-92-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.