sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number ACR107700010
Price (MYR)
451.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
451.00
Add to cart
Chemical Identifiers
CAS718-64-9
IUPAC Name1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-ol
Molecular FormulaC9H6F6O
InChI KeyIZPIZCAYJQCTNG-UHFFFAOYSA-N
SMILESOC(C1=CC=CC=C1)(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4135-1.4175 @ 20?C
Assay (GC)≥98.0%